Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013481.1 | NP_038509.1 | 14 | 1957 | ||
GenBank mRNA | AK079627.1 | - | 14 | 1937 | ||
AK129058.1 | - | 14 | 1930 | |||
AK144095.1 | - | 14 | 1969 | |||
AK167110.1 | - | 14 | 1940 | |||
AK167225.1 | - | 14 | 1941 | |||
AK167829.1 | - | 14 | 1935 | |||
AK167878.1 | - | 14 | 1945 | |||
AK168127.1 | - | 14 | 1938 | |||
BC012693.1 | - | 14 | 1931 | 4193123 | ||
U77415.1 | - | 14 | 1957 |