Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013481.1 | NP_038509.1 | 6 | 757 | ||
GenBank mRNA | AK079627.1 | - | 6 | 737 | ||
AK144095.1 | - | 6 | 768 | |||
AK167110.1 | - | 6 | 740 | |||
AK167225.1 | - | 6 | 741 | |||
AK167829.1 | - | 6 | 735 | |||
AK167878.1 | - | 6 | 745 | |||
AK168127.1 | - | 6 | 738 | |||
BC012693.1 | - | 6 | 731 | 4193123 | ||
U77415.1 | - | 6 | 757 |