Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009767.2 | NP_033897.1 | 3 | 612 | ||
GenBank mRNA | AK013993.1 | - | 3 | 512 | ||
AK034427.1 | - | 3 | 585 | |||
AK135974.1 | - | 3 | 564 | |||
BC132322.1 | - | 3 | 531 | 40130599 | ||
BC138730.1 | - | 3 | 531 | 8861752 |