Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007568.5 | NP_031594.1 | 4, 5 | 704 | ||
GenBank mRNA | AK050206.1 | - | 4, 5 | 520 | ||
AK076272.1 | - | 4, 5 | 624 | |||
AK164633.1 | - | 4, 5 | 705 | |||
BC119152.1 | - | 4, 5 | 570 | 8733901 | ||
BC119154.1 | - | 4, 5 | 570 | 8733903 | ||
BC144906.1 | - | 3, 4 | 452 | |||
L08394.1 | - | 4, 5 | 543 |