Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007568.5 | NP_031594.1 | 3 | 457 | ||
GenBank mRNA | AK046658.1 | - | 3 | 287 | ||
AK050206.1 | - | 3 | 273 | |||
AK076272.1 | - | 3 | 377 | |||
AK164633.1 | - | 3 | 458 | |||
BC119152.1 | - | 3 | 323 | 8733901 | ||
BC119154.1 | - | 3 | 323 | 8733903 | ||
L08394.1 | - | 3 | 296 |