Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bub1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113179.1 | NP_001106650.1 | 8 | 845 | ||
NM_009772.2 | NP_033902.2 | 8 | 842 | |||
GenBank mRNA | AF002823.1 | - | 8 | 788 | ||
AK021258.1 | - | 8 | 833 | |||
AK042932.1 | - | 8 | 843 | |||
AK144213.1 | - | 8 | 854 | |||
BC029116.1 | - | 8 | 811 | 3671932 | ||
U89795.1 | - | 8 | 860 |