Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009775.4 | NP_033905.3 | 4 | 510 | ||
GenBank mRNA | AA472650.1 | - | 4 | 507 | ||
AK149743.1 | - | 4 | 485 | |||
AK150516.1 | - | 4 | 482 | |||
AK151421.1 | - | 4 | 484 | |||
AK151569.1 | - | 4 | 491 | |||
AK151824.1 | - | 4 | 491 | |||
AK152082.1 | - | 4 | 482 | |||
AK152827.1 | - | 4 | 482 | |||
AK152860.1 | - | 4 | 485 | |||
AK153159.1 | - | 4 | 484 | |||
AK153273.1 | - | 4 | 494 | |||
AK156489.1 | - | 4 | 485 | |||
AK165450.1 | - | 4 | 484 | |||
AK165822.1 | - | 4 | 491 | |||
AK166125.1 | - | 4 | 491 | |||
AK172384.1 | - | 4 | 485 | |||
BC002055.1 | - | 4 | 472 | 3493196 | ||
D21207.1 | - | 4 | 474 | |||
L17306.1 | - | 3 | 439 |