Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC043338.1 | - | 24 | 3112 | 5134713 | |
DQ408205.1 | - | 1 | 55 | |||
EU868829.1 | - | 1 | 55 | |||
HM856604.1 | - | 1 | 76 | |||
K02782.1 | - | 24 | 3126 |