Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2H4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001517.4 | NP_001508.1 | 6, 7 | 770 | ||
GenBank mRNA | AK222607.1 | - | 6, 7 | 674 | ||
AK298058.1 | - | 6, 7 | 818 | |||
AK300239.1 | - | 5, 6 | 623 | |||
AK309869.1 | - | 6, 7 | 784 | |||
BC004935.1 | - | 6, 7 | 742 | 3606718 | ||
BC016302.2 | - | 6, 7 | 742 | 3830902 | ||
BT007321.1 | - | 5, 6 | 563 | |||
Y07595.1 | - | 6, 7 | 690 |