Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hyou1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021395.4 | NP_067370.3 | 3 | 285 | ||
GenBank mRNA | AF228709.1 | - | 2 | 172 | ||
AK128937.1 | - | 3 | 232 | |||
AK145857.1 | - | 3 | 233 | |||
AK155721.1 | - | 3 | 233 | |||
AK157949.1 | - | 3 | 203 | |||
AK171769.1 | - | 3 | 233 | |||
BC019785.1 | - | 2 | 191 | 5151731 | ||
BC050107.1 | - | 2 | 190 | 30012694 | ||
BC058255.1 | - | 3 | 218 |