Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cab39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133781.4 | NP_598542.3 | 8 | 1149 | ||
GenBank mRNA | AK144665.1 | - | 9 | 982 | ||
AK199827.1 | - | 4 | 301 | |||
BC020041.1 | - | 8 | 811 | 3990872 | ||
BC029053.1 | - | 4 | 323 | |||
S51858.1 | - | 8 | 886 |