Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacna1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB086123.1 | - | 6 | 875 | ||
AJ437291.1 | - | 6 | 857 | |||
AJ437292.1 | - | 6 | 978 | |||
AK147415.1 | - | 6 | 1353 |