Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307936.1 | NP_001294865.1 | 9 | 1140 | ||
NM_018976.4 | NP_061849.2 | 10 | 1256 | |||
GenBank mRNA | AB037803.2 | - | 9 | 816 | ||
AF259799.1 | - | 9 | 836 | |||
AF298897.1 | - | 10 | 1167 | |||
AJ344099.1 | - | 10 | 1168 | |||
AK172784.1 | - | 3 | 263 | |||
AK291644.1 | - | 10 | 960 | |||
AK293858.1 | - | 3 | 371 | |||
BC040342.1 | - | 10 | 1158 | 3874551 | ||
CR457267.1 | - | 6 | 516 |