Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacna2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110843.1 | NP_001104313.1 | 39 | 3576 | ||
NM_001110844.1 | NP_001104314.1 | 39 | 3540 | |||
NM_001110845.1 | NP_001104315.1 | 39 | 3525 | |||
NM_001110846.1 | NP_001104316.1 | 38 | 3504 | |||
NM_009784.2 | NP_033914.1 | 38 | 3519 | |||
GenBank mRNA | AK014825.1 | - | 34 | 2809 | ||
AK053484.1 | - | 21 | 1669 | |||
BC115871.1 | - | 40 | 3300 | 40061614 | ||
U73483.1 | - | 39 | 3483 | |||
U73484.1 | - | 39 | 3447 | |||
U73485.1 | - | 39 | 3432 | |||
U73486.1 | - | 38 | 3411 | |||
U73487.1 | - | 38 | 3426 |