Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037099.2 | NP_001032176.1 | 11, 12 | 1305 | ||
NM_001285426.1 | NP_001272355.1 | 11, 12 | 1089 | |||
NM_001285427.1 | NP_001272356.1 | 11, 12 | 1385 | |||
NM_001285428.1 | NP_001272357.1 | 10, 11 | 1014 | |||
NM_146123.3 | NP_666235.1 | 10, 11 | 1225 | |||
GenBank mRNA | AF039417.1 | - | 1, 2 | 82 | ||
AK032311.1 | - | 11, 12 | 1528 | |||
AK038633.1 | - | 11, 12 | 1341 | |||
AK043850.1 | - | 10, 11 | 1168 | |||
AK079616.1 | - | 10, 11 | 1175 | |||
AK147338.1 | - | 11, 12 | 1279 | |||
AK158606.1 | - | 10, 11 | 1016 | |||
BC026479.1 | - | 10, 11 | 1011 | 4501980 |