Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacybp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009786.2 | NP_033916.1 | 2 | 444 | ||
GenBank mRNA | AK013924.1 | - | 2 | 375 | ||
AK089043.1 | - | 2 | 298 | |||
AK145013.1 | - | 2 | 411 | |||
BC025948.1 | - | 2 | 329 | 5150537 | ||
U97327.1 | - | 2 | 274 |