Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007585.3 | NP_031611.1 | 8 | 630 | ||
GenBank mRNA | AK012563.1 | - | 9 | 632 | ||
AK088645.1 | - | 8 | 619 | |||
AK146096.1 | - | 8 | 618 | |||
AK150586.1 | - | 8 | 618 | |||
AK152561.1 | - | 8 | 618 | |||
AK160746.1 | - | 8 | 618 | |||
AK164507.1 | - | 8 | 623 | |||
AK165888.1 | - | 8 | 618 | |||
AK167894.1 | - | 8 | 617 | |||
AK168083.1 | - | 8 | 618 | |||
BC003327.1 | - | 8 | 630 | 3156634 | ||
BC004659.1 | - | 4 | 1526 | 3497991 | ||
BC005763.1 | - | 8 | 604 | 2655513 | ||
D10024.1 | - | 7 | 563 | |||
M14044.1 | - | 7 | 563 |