Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007586.1 | NP_031612.1 | 1, 2 | 155 | ||
GenBank mRNA | AK032171.1 | - | 161 | |||
BC017646.1 | - | 1, 2 | 155 | 4527074 |