Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009790.4 | NP_033920.1 | 3 | 328 | ||
GenBank mRNA | AK004673.1 | - | 3 | 342 | ||
AK013695.1 | - | 3 | 337 | |||
AK088141.1 | - | 3 | 338 | |||
AK145841.1 | - | 3 | 315 | |||
AK150288.1 | - | 3 | 338 | |||
AK150978.1 | - | 3 | 343 | |||
AK151001.1 | - | 3 | 338 | |||
AK151552.1 | - | 3 | 338 | |||
AK151784.1 | - | 3 | 343 | |||
AK151923.1 | - | 3 | 341 | |||
AK151992.1 | - | 3 | 343 | |||
AK152148.1 | - | 3 | 341 | |||
AK152715.1 | - | 3 | 337 | |||
AK152719.1 | - | 3 | 338 | |||
AK152850.1 | - | 3 | 341 | |||
AK152897.1 | - | 3 | 338 | |||
AK153004.1 | - | 3 | 337 | |||
AK153348.1 | - | 3 | 341 | |||
AK153426.1 | - | 3 | 341 | |||
AK153546.1 | - | 3 | 341 | |||
AK159762.1 | - | 3 | 344 | |||
AK160057.1 | - | 3 | 344 | |||
AK160508.1 | - | 3 | 343 | |||
AK161302.1 | - | 3 | 299 | |||
AK162314.1 | - | 3 | 345 | |||
AK166308.1 | - | 3 | 338 | |||
AK167353.1 | - | 3 | 343 | |||
AK168002.1 | - | 3 | 341 | |||
AK168241.1 | - | 3 | 297 | |||
AK168663.1 | - | 3 | 337 | |||
AK168803.1 | - | 3 | 341 | |||
AK169027.1 | - | 3 | 343 | |||
AK169055.1 | - | 3 | 341 | |||
AK185360.1 | - | 1 | 44 | |||
BC054805.1 | - | 3 | 328 | 6838207 | ||
M19381.1 | - | 3 | 341 | |||
X61432.1 | - | 3 | 181 |