Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007589.5 | NP_031615.1 | 5 | 519 | ||
GenBank mRNA | AK002974.1 | - | 480 | |||
AK012247.1 | - | 5 | 447 | |||
AK012564.1 | - | 5 | 448 | |||
AK013068.1 | - | 5 | 441 | |||
AK152303.1 | - | 5 | 451 | |||
AK160636.1 | - | 5 | 454 | |||
AK161268.1 | - | 5 | 447 | |||
AK161984.1 | - | 5 | 448 | |||
AK168741.1 | - | 5 | 448 | |||
BC010730.1 | - | 3 | 224 | |||
BC021347.1 | - | 5 | 450 | 4006686 | ||
BC051444.1 | - | 5 | 443 | 30072702 | ||
BC100301.1 | - | 5 | 434 | 30526959 | ||
L31642.1 | - | 5 | 454 |