Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174054.1 | NP_001167525.1 | 16 | 1318 | ||
NM_007595.5 | NP_031621.3 | 16 | 1357 | |||
GenBank mRNA | AK046454.1 | - | 16 | 1358 | ||
AK163698.1 | - | 16 | 1174 | |||
BC024803.1 | - | 16 | 1134 | |||
X63615.1 | - | 16 | 1172 |