Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301067.1 | NP_001287996.1 | 11 | 1419 | ||
NM_007598.4 | NP_031624.2 | 11 | 1403 | |||
GenBank mRNA | AK053351.1 | - | 11 | 1227 | ||
AK128889.1 | - | 11 | 1225 | |||
AK137222.1 | - | 11 | 1208 | |||
AK144094.1 | - | 11 | 1211 | |||
AK154002.1 | - | 11 | 1423 | |||
AK156556.1 | - | 11 | 1237 | |||
AK158150.1 | - | 10 | 2067 | |||
AK165565.1 | - | 11 | 1206 | |||
AK169652.1 | - | 11 | 1198 | |||
AK170901.1 | - | 11 | 1214 | |||
AK171388.1 | - | 11 | 1221 | |||
BC005446.1 | - | 10 | 1202 | 3600514 | ||
BC005472.1 | - | 10 | 1195 | 3590890 | ||
L12367.1 | - | 11 | 1229 |