Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018976.4 | NP_061849.2 | 4 | 736 | ||
GenBank mRNA | AB037803.2 | - | 3 | 296 | ||
AF259799.1 | - | 3 | 316 | |||
AF298897.1 | - | 4 | 647 | |||
AJ344099.1 | - | 4 | 648 | |||
AK291644.1 | - | 4 | 440 | |||
BC040342.1 | - | 4 | 638 | 3874551 |