Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009794.3 | NP_033924.2 | 12 | 1475 | ||
GenBank mRNA | AF015038.1 | - | 12 | 1421 | ||
AK150695.1 | - | 12 | 1449 | |||
AK152094.1 | - | 12 | 1449 | |||
AK152790.1 | - | 12 | 1452 | |||
AK152994.1 | - | 12 | 1452 | |||
AK154855.1 | - | 12 | 1489 | |||
AK181353.1 | - | 3 | 101 | |||
AK207878.1 | - | 3 | 101 | |||
BC054726.1 | - | 12 | 1475 | 5703563 | ||
D38117.1 | - | 12 | 1516 | |||
Y10139.1 | - | 12 | 1357 |