Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009794.3 | NP_033924.2 | 17 | 1939 | ||
GenBank mRNA | AF015038.1 | - | 17 | 1885 | ||
AK052141.1 | - | 15 | 1527 | |||
AK150695.1 | - | 17 | 1913 | |||
AK152094.1 | - | 17 | 1913 | |||
AK152790.1 | - | 17 | 1916 | |||
AK152994.1 | - | 17 | 1916 | |||
AK154855.1 | - | 17 | 1953 | |||
AK183150.1 | - | 6 | 375 | |||
AK183766.1 | - | 6 | 339 | |||
AK195491.1 | - | 6 | 383 | |||
AK199295.1 | - | 6 | 375 | |||
BC054726.1 | - | 17 | 1939 | 5703563 | ||
D38117.1 | - | 17 | 1980 | |||
Y10139.1 | - | 17 | 1821 |