Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009794.3 | NP_033924.2 | 4, 5 | 698 | ||
GenBank mRNA | AF015038.1 | - | 4, 5 | 644 | ||
AK052141.1 | - | 4, 5 | 673 | |||
AK078787.1 | - | 1, 2 | 509 | |||
AK150695.1 | - | 4, 5 | 672 | |||
AK152094.1 | - | 4, 5 | 672 | |||
AK152790.1 | - | 4, 5 | 672 | |||
AK152994.1 | - | 4, 5 | 672 | |||
AK154855.1 | - | 4, 5 | 712 | |||
AK184788.1 | - | 95 | ||||
BC054726.1 | - | 4, 5 | 698 | 5703563 | ||
D38117.1 | - | 4, 5 | 739 | |||
Y10139.1 | - | 4, 5 | 580 |