Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083957.1 | NP_001077426.1 | 3 | 410 | ||
NM_009799.4 | NP_033929.2 | 4 | 422 | |||
GenBank mRNA | AK003066.1 | - | 4 | 423 | ||
AK033719.1 | - | 3 | 411 | |||
AK146372.1 | - | 4 | 423 | |||
AK162331.1 | - | 3 | 410 | |||
AK167969.1 | - | 4 | 422 | |||
AK172076.1 | - | 4 | 423 | |||
BC011223.1 | - | 3 | 394 | 4208180 | ||
BC110681.1 | - | 3 | 392 | 4984216 | ||
BC132432.1 | - | 4 | 378 | 40130709 | ||
BC132434.1 | - | 4 | 378 | 40130711 | ||
M32452.1 | - | 3 | 379 |