Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083957.1 | NP_001077426.1 | 2, 3 | 309 | ||
NM_009799.4 | NP_033929.2 | 3, 4 | 321 | |||
GenBank mRNA | AK003066.1 | - | 3, 4 | 322 | ||
AK033719.1 | - | 2, 3 | 310 | |||
AK146372.1 | - | 3, 4 | 322 | |||
AK162331.1 | - | 2, 3 | 309 | |||
AK167969.1 | - | 3, 4 | 321 | |||
AK172076.1 | - | 3, 4 | 322 | |||
BB842334.1 | - | 3, 4 | 322 | |||
BC011223.1 | - | 2, 3 | 293 | 4208180 | ||
BC110681.1 | - | 2, 3 | 291 | 4984216 | ||
BC132432.1 | - | 3, 4 | 277 | 40130709 | ||
BC132434.1 | - | 3, 4 | 277 | 40130711 | ||
M32452.1 | - | 2, 3 | 278 |