Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009801.4 | NP_033931.4 | 2 | 476 | ||
GenBank mRNA | AK002333.1 | - | 2 | 211 | ||
AK146015.1 | - | 2 | 212 | |||
BC044806.1 | - | 2 | 196 | |||
BC055291.1 | - | 2 | 196 | 6479187 | ||
BU946040.1 | - | 2 | 478 | |||
K00811.1 | - | 2 | 177 |