Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301164.1 | NP_001288093.1 | 5 | 986 | ||
NM_001301165.1 | NP_001288094.1 | 5 | 554 | |||
NM_053070.3 | NP_444300.1 | 5 | 581 | |||
GenBank mRNA | AF291660.1 | - | 4 | 476 | ||
AK090390.1 | - | 5 | 585 | |||
AK144275.1 | - | 5 | 555 | |||
AY075021.1 | - | 5 | 582 | |||
BC038270.1 | - | 5 | 961 | |||
BC094913.1 | - | 5 | 569 | 30016726 |