Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000409.4 | NP_000400.2 | 4 | 891 | ||
NM_001319061.1 | NP_001305990.1 | 4 | 1262 | |||
NM_001319062.1 | NP_001305991.1 | 3 | 715 | |||
XM_006715073.3 | XP_006715136.1 | 3 | 104 | |||
XM_011514537.2 | XP_011512839.1 | 4 | 1152 | |||
XM_011514539.1 | XP_011512841.1 | 4 | 754 | |||
GenBank mRNA | AK125780.1 | - | 4 | 1193 | ||
AK313865.1 | - | 3 | 686 | |||
BC031663.1 | - | 4 | 832 | 5172128 | ||
L36859.1 | - | 2 | 240 |