Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1i3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243062.1 | NP_001229991.1 | 5 | 661 | ||
NM_001243063.1 | NP_001229992.1 | 5 | 661 | |||
NM_009803.5 | NP_033933.2 | 5 | 661 | |||
GenBank mRNA | AK133515.1 | - | 5 | 662 | ||
AK149381.1 | - | 5 | 613 | |||
AK149430.1 | - | 372 | ||||
BC089541.1 | - | 5 | 610 | 30315989 |