Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080126.1 | NP_001073595.1 | 9 | 1567 | ||
NM_001277926.1 | NP_001264855.1 | 10 | 1683 | |||
NM_009812.2 | NP_033942.1 | 9 | 1740 | |||
GenBank mRNA | AF067834.1 | - | 9 | 1620 | ||
AJ000641.1 | - | 8 | 1186 | |||
AJ007749.1 | - | 9 | 1393 | |||
AK146758.1 | - | 9 | 1740 | |||
AK153340.1 | - | 9 | 1449 | |||
AK156135.1 | - | 9 | 1456 | |||
AK168812.1 | - | 9 | 1740 | |||
BC006737.1 | - | 9 | 1682 | 3964119 | ||
BC049955.1 | - | 9 | 1574 | 4208824 | ||
CT010166.1 | - | 8 | 1347 |
Gene Symbol: Gm20257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079804.1 | - | 2 | 86 |