Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cast | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301153.1 | NP_001288082.1 | 19 | 1630 | ||
NM_001301155.1 | NP_001288084.1 | 17 | 1361 | |||
NM_001301156.1 | NP_001288085.1 | 16 | 1304 | |||
NM_001301157.1 | NP_001288086.1 | 16 | 1304 | |||
NM_001301158.1 | NP_001288087.1 | 4 | 438 | |||
NM_001301160.1 | NP_001288089.1 | 4 | 393 | |||
NM_001301181.1 | NP_001288110.1 | 4 | 438 | |||
NM_009817.4 | NP_033947.1 | 18 | 1390 | |||
GenBank mRNA | AB026997.1 | - | 19 | 1629 | ||
AF190151.1 | - | 4 | 336 | |||
AF190152.1 | - | 4 | 381 | |||
AK006253.1 | - | 4 | 440 | |||
AK028841.1 | - | 18 | 1370 | |||
AK028843.1 | - | 18 | 1352 | |||
AK029293.1 | - | 17 | 1277 | |||
AK036576.1 | - | 16 | 1175 | |||
AK083310.1 | - | 16 | 1207 | |||
AK083395.1 | - | 16 | 1196 | |||
AK089108.1 | - | 16 | 1202 | |||
AK161284.1 | - | 4 | 366 | |||
BC010603.1 | - | 16 | 1158 | 3710078 |