Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161456.1 | NP_001154928.1 | 4 | 636 | ||
NM_001161458.1 | NP_001154930.1 | 5 | 753 | |||
NM_022309.4 | NP_071704.3 | 5 | 753 | |||
GenBank mRNA | AK089094.1 | - | 5 | 754 | ||
AK134449.1 | - | 4 | 321 | |||
AK198845.1 | - | 203 | ||||
BC006763.1 | - | 5 | 521 | 3481800 | ||
BC026749.1 | - | 5 | 563 | 4016060 | ||
BC040752.1 | - | 5 | 552 | 4016060 | ||
D14571.1 | - | 5 | 683 | |||
D14572.1 | - | 5 | 683 | |||
L03279.1 | - | 5 | 532 | |||
L03305.1 | - | 4 | 374 | |||
L03306.1 | - | 451 |