Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019626.3 | NP_062600.2 | 2 | 689 | ||
GenBank mRNA | AK158818.1 | - | 2 | 690 | ||
AK158885.1 | - | 2 | 707 | |||
AK161846.1 | - | 2 | 288 | |||
BC051956.1 | - | 2 | 304 | |||
BC055730.1 | - | 2 | 393 | 6404555 | ||
BC132120.1 | - | 3 | 372 | 40130397 | ||
BC132122.1 | - | 3 | 438 | 40130399 |