Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007624.3 | NP_031650.3 | 3, 4 | 323 | ||
GenBank mRNA | AK002910.1 | - | 3, 4 | 298 | ||
AK083957.1 | - | 3, 4 | 238 | |||
AK146376.1 | - | 3, 4 | 273 | |||
BC003840.1 | - | 3, 4 | 249 | |||
BC018354.1 | - | 2, 3 | 205 | 3990224 | ||
BC059831.1 | - | 3, 4 | 226 | 5717629 | ||
BC108370.1 | - | 3, 4 | 250 | 30728322 | ||
BC110376.1 | - | 3, 4 | 280 | 3467208 | ||
CJ093936.1 | - | 3, 4 | 324 | |||
X56683.1 | - | 1, 2 | 140 |