Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCY1B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000857.3 | NP_000848.1 | 5, 6 | 657 | ||
NM_001291951.1 | NP_001278880.1 | 6, 7 | 723 | |||
NM_001291952.1 | NP_001278881.1 | 6, 7 | 809 | |||
NM_001291953.1 | NP_001278882.1 | 6, 7 | 843 | |||
NM_001291954.1 | NP_001278883.1 | 5, 6 | 657 | |||
NM_001291955.1 | NP_001278884.1 | 4, 5 | 583 | |||
XM_011531901.1 | XP_011530203.1 | 5, 6 | 571 | |||
XM_017008130.1 | XP_016863619.1 | 5, 6 | 848 | |||
XM_017008131.1 | XP_016863620.1 | 4, 5 | 559 | |||
XM_017008132.1 | XP_016863621.1 | 5, 6 | 848 | |||
XM_017008133.1 | XP_016863622.1 | 4, 5 | 635 | |||
GenBank mRNA | AF020340.1 | - | 5, 6 | 540 | ||
AK296680.1 | - | 6, 7 | 756 | |||
AK299916.1 | - | 5, 6 | 569 | |||
AK307838.1 | - | 6, 7 | 785 | |||
AK315199.1 | - | 5, 6 | 610 | |||
AK315944.1 | - | 4, 5 | 564 | |||
BC047620.1 | - | 5, 6 | 571 | 5264160 | ||
X66533.1 | - | 5, 6 | 577 |