Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284508.1 | NP_001271437.1 | 2 | 301 | ||
NM_031161.4 | NP_112438.1 | 2 | 301 | |||
GenBank mRNA | AK002677.1 | - | 2 | 269 | ||
BC028487.1 | - | 2 | 246 | 1400830 | ||
BY127153.1 | - | 2 | 270 | |||
BY268487.1 | - | 2 | 302 | |||
M11739.1 | - | 1 | 40 |