Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cckbr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF019371.1 | - | 3 | 638 | ||
AK028325.1 | - | 3 | 848 | |||
AK039721.1 | - | 847 | ||||
AK053307.1 | - | 3 | 708 | |||
BC103530.1 | - | 3 | 818 | 40039482 | ||
BC103531.1 | - | 3 | 818 | 40039474 | ||
BC103537.1 | - | 4 | 767 | |||
BC103538.1 | - | 3 | 818 | 40039475 |