Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009831.2 | NP_033961.1 | 2 | 532 | ||
GenBank mRNA | AK128957.1 | - | 2 | 520 | ||
AK136645.1 | - | 2 | 270 | |||
AK143114.1 | - | 2 | 523 | |||
AK147026.1 | - | 2 | 520 | |||
AK153392.1 | - | 2 | 464 | |||
AK159429.1 | - | 2 | 462 | |||
AK159831.1 | - | 2 | 467 | |||
AK160411.1 | - | 2 | 507 | |||
AK167052.1 | - | 2 | 464 | |||
AK167486.1 | - | 2 | 459 | |||
AK170397.1 | - | 2 | 520 | |||
BC005534.1 | - | 1 | 182 | |||
BC006609.1 | - | 2 | 416 | |||
L49507.1 | - | 2 | 475 | |||
Z37110.1 | - | 2 | 475 |