Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccni | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017367.3 | NP_059063.2 | 3 | 844 | ||
GenBank mRNA | AF005886.2 | - | 3 | 496 | ||
AK050465.1 | - | 3 | 845 | |||
AK079476.1 | - | 3 | 785 | |||
AK152614.1 | - | 3 | 801 | |||
AK208767.1 | - | 1 | 117 | |||
AK212895.1 | - | 3 | 302 | |||
BC003290.1 | - | 3 | 377 | 3583418 | ||
BC029328.1 | - | 3 | 377 |