Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190333.1 | NP_001177262.1 | 3 | 913 | ||
NM_001190334.1 | NP_001177263.1 | 4 | 785 | |||
NM_001190335.1 | NP_001177264.1 | 3 | 694 | |||
NM_001190336.1 | NP_001177265.1 | 3 | 670 | |||
NM_001190337.1 | NP_001177266.1 | 2 | 579 | |||
NM_001190338.1 | NP_001177267.1 | 2 | 666 | |||
NM_009835.4 | NP_033965.1 | 6 | 1115 | |||
GenBank mRNA | AB009369.1 | - | 2 | 669 | ||
AB016031.1 | - | 3 | 744 | |||
AK089712.1 | - | 6 | 1116 | |||
BC100756.1 | - | 4 | 781 | 40046797 | ||
BC100757.1 | - | 3 | 690 | 40046796 | ||
BC100758.1 | - | 3 | 666 | 40046794 | ||
BC105669.1 | - | 2 | 572 | 40046795 |