Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287593.1 | NP_001274522.1 | 5 | 895 | ||
NM_001287594.1 | NP_001274523.1 | 5 | 435 | |||
NM_001288.4 | NP_001279.2 | 4 | 586 | |||
GenBank mRNA | AF034607.1 | - | 4 | 591 | ||
AF109197.1 | - | 4 | 376 | |||
AK129773.1 | - | 4 | 331 | |||
AK223379.1 | - | 4 | 559 | |||
AK314014.1 | - | 4 | 331 | |||
BC064527.1 | - | 5 | 895 | 5585323 | ||
BC095469.1 | - | 4 | 521 | 30557606 | ||
BM562536.1 | - | 589 | ||||
CB124154.1 | - | 2 | 123 | |||
CR542071.1 | - | 4 | 322 | |||
CX783058.1 | - | 5 | 433 | |||
HY057217.1 | - | 5 | 436 | |||
U93205.1 | - | 4 | 540 | |||
X87689.1 | - | 4 | 526 |