Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001043317.2 | NP_001036782.1 | 6, 7 | 1118 | ||
NM_009845.3 | NP_033975.3 | 4, 5 | 993 | |||
GenBank mRNA | AF102134.1 | - | 3, 4 | 762 | ||
AF115401.1 | - | 3, 4 | 762 | |||
AK143835.1 | - | 6, 7 | 1056 | |||
AK171589.1 | - | 3, 4 | 799 | |||
BC046957.1 | - | 6, 7 | 1044 | |||
L16928.1 | - | 5, 6 | 1241 |