Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001043317.2 | NP_001036782.1 | 15 | 2818 | ||
NM_009845.3 | NP_033975.3 | 13 | 2693 | |||
GenBank mRNA | AF102134.1 | - | 12 | 2462 | ||
AF115400.1 | - | 12 | 2462 | |||
AF115401.1 | - | 12 | 2462 | |||
AK041230.1 | - | 15 | 2825 | |||
AK143835.1 | - | 15 | 2756 | |||
AK156735.1 | - | 13 | 2692 | |||
AK171589.1 | - | 12 | 2499 | |||
AK172301.1 | - | 13 | 2649 | |||
BC046957.1 | - | 15 | 2763 | |||
BC150807.1 | - | 14 | 2777 | 9087718 | ||
L02844.1 | - | 14 | 2964 | |||
L16928.1 | - | 14 | 2941 |