Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159555.1 | NP_001153027.1 | 14 | 1833 | ||
NM_001159556.1 | NP_001153028.1 | 14 | 1693 | |||
NM_001159557.1 | NP_001153029.1 | 15 | 1744 | |||
NM_001159558.1 | NP_001153030.1 | 15 | 1860 | |||
NM_007643.4 | NP_031669.3 | 14 | 1836 | |||
GenBank mRNA | AK004192.1 | - | 2 | 155 | ||
AK052825.1 | - | 14 | 1637 | |||
AK128908.1 | - | 15 | 1637 | |||
AK150648.1 | - | 14 | 1657 | |||
AK150865.1 | - | 10 | 1029 | |||
AK151355.1 | - | 14 | 1672 | |||
AK152642.1 | - | 9 | 828 | |||
AK152904.1 | - | 12 | 1343 | |||
AK153095.1 | - | 9 | 946 | |||
AK172162.1 | - | 15 | 1682 | |||
BC010262.1 | - | 14 | 1645 | 3481681 | ||
L23108.1 | - | 14 | 1654 |