Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159555.1 | NP_001153027.1 | 11 | 1571 | ||
NM_001159556.1 | NP_001153028.1 | 11 | 1431 | |||
NM_001159557.1 | NP_001153029.1 | 12 | 1482 | |||
NM_001159558.1 | NP_001153030.1 | 12 | 1598 | |||
NM_007643.4 | NP_031669.3 | 11 | 1574 | |||
GenBank mRNA | AK052825.1 | - | 11 | 1375 | ||
AK128908.1 | - | 12 | 1375 | |||
AK150865.1 | - | 7 | 769 | |||
AK151355.1 | - | 11 | 1410 | |||
AK152642.1 | - | 6 | 566 | |||
AK152904.1 | - | 9 | 1081 | |||
AK153095.1 | - | 6 | 684 | |||
AK172162.1 | - | 12 | 1420 | |||
BC010262.1 | - | 11 | 1383 | 3481681 | ||
HQ171908.1 | - | 2 | 145 | |||
L23108.1 | - | 11 | 1392 |