Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159555.1 | NP_001153027.1 | 10 | 1425 | ||
NM_001159556.1 | NP_001153028.1 | 10 | 1285 | |||
NM_001159557.1 | NP_001153029.1 | 11 | 1336 | |||
NM_001159558.1 | NP_001153030.1 | 11 | 1452 | |||
NM_007643.4 | NP_031669.3 | 10 | 1428 | |||
GenBank mRNA | AK052825.1 | - | 10 | 1229 | ||
AK128908.1 | - | 11 | 1229 | |||
AK150648.1 | - | 10 | 1249 | |||
AK150865.1 | - | 6 | 623 | |||
AK151355.1 | - | 10 | 1264 | |||
AK152642.1 | - | 5 | 420 | |||
AK152904.1 | - | 8 | 935 | |||
AK153095.1 | - | 5 | 538 | |||
AK172162.1 | - | 11 | 1274 | |||
BC010262.1 | - | 10 | 1237 | 3481681 | ||
L23108.1 | - | 10 | 1246 |