Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290802.1 | NP_001277731.1 | 8 | 887 | ||
NM_001290804.1 | NP_001277733.1 | 7 | 812 | |||
NM_007645.4 | NP_031671.1 | 7 | 836 | |||
GenBank mRNA | AK154250.1 | - | 7 | 876 | ||
AK154466.1 | - | 8 | 857 | |||
AK155663.1 | - | 6 | 1452 | |||
AK155966.1 | - | 7 | 801 | |||
BC019402.1 | - | 7 | 873 | 4013070 | ||
BY548149.1 | - | 1 | 48 | |||
CT010217.1 | - | 7 | 725 |